CAS RN 593-50-0
CAS RN 593-50-0
IUPAC Name:
triacontan-1-ol
Synonyms:
triacontanol aluminum salt 1-triacontanol myricyl alcohol melissyl alcohol
Molecular Weight (g/mol):
438.83
Molecular Formula:
C30H62O
InChi Key:
REZQBEBOWJAQKS-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO
1
–
2
of
2
results
2
1-Triacontanol (>90% purity), TRC
CAS: 593-50-0 Molecular Formula: C30 H62 O Molecular Weight (g/mol): 438.81 Synonym: 1-Triacontanol,1-Triacontanol (6CI, 7CI, 8CI, 9CI, ACI),1-Hydroxytriacontane,Melissyl alcohol,Miraculan,Myricyl alcohol,n-Triacontanol,NSC 402492,NSC 405588,Nutron,Prosopol,Tomatex,Triacontanol,Triacontyl alcohol,Ultria,Well-Bloom IUPAC Name: triacontan-1-ol SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO