missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Melting Point | 37.0°C to 40.0°C |
| Color | White |
| Boiling Point | 289.0°C |
| Flash Point | 145°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | CH3(CH2)13OH |
| Show More |
Safety and Handling
Product Identifier
- 1-Tetradecanol
- Warning
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to an approved waste disposal plant.
- MIXTURE LIST-Contain: Myristic alcohol, Lauryl alcohol
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?