missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecanol, 97+%

Product Code. 11414150
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414150 100 g 100g
11424187 500 g 500g
11434187 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414150 Supplier Thermo Scientific Alfa Aesar Supplier No. A19638.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 112-72-1 | C14H30O | 214.39 g/mol

1-Tetradecanol is used as an ingredient in cosmetics such as cold creams. It is an active intermediate in the chemical synthesis of sulfated alcohol. It is also employed in the fabrication of temperature-regulated drug release system based on phase-change materials. It plays a vital role in filling the hollow interiors of gold nanocages in the fabrication of new theranostic system, which has unique feature of photoacoustic imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tetradecanol is used as an ingredient in cosmetics such as cold creams. It is an active intermediate in the chemical synthesis of sulfated alcohol. It is also employed in the fabrication of temperature-regulated drug release system based on phase-change materials. It plays a vital role in filling the hollow interiors of gold nanocages in the fabrication of new theranostic system, which has unique feature of photoacoustic imaging.

Solubility
Insoluble in water. Soluble in diethyl ether. Slightly soluble in ethanol.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-72-1
Molecular Formula C14H30O
Molecular Weight (g/mol) 214.39
MDL Number MFCD00004757
InChI Key HLZKNKRTKFSKGZ-UHFFFAOYSA-N
Synonym 1-tetradecanol, myristyl alcohol, tetradecanol, tetradecyl alcohol, n-tetradecanol, myristic alcohol, n-tetradecyl alcohol, lanette k, loxanol v, lanette wax ks
PubChem CID 8209
ChEBI CHEBI:77417
IUPAC Name tetradecan-1-ol
SMILES CCCCCCCCCCCCCCO

Specifications

Melting Point 37°C to 40°C
Density 0.823
Boiling Point 289°C
Flash Point 141°C (285°F)
Quantity 100 g
Beilstein 1742652
Merck Index 14,6334
Solubility Information Insoluble in water. Soluble in diethyl ether. Slightly soluble in ethanol.
Formula Weight 214.39
Percent Purity ≥97%
Chemical Name or Material 1-Tetradecanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.