missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl-3-Methyl 3-Hydroxypyrrolidine-1,3-Dicarboxylate, TRC

Product Code. 30416921
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30416921 500 mg
30419429 100 mg
30418498 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30416921 Supplier Toronto Research Chemicals Supplier No. TRCT206338500MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 942190-61-6
Molecular Formula C11H19NO5
Molecular Weight (g/mol) 245.27
Synonym 1-(1,1-Dimethylethyl) 3-methyl 3-hydroxy-1,3-pyrrolidinedicarboxylate
IUPAC Name 1-O-tert-butyl 3-O-methyl 3-hydroxypyrrolidine-1,3-dicarboxylate
SMILES COC(=O)C1(O)CCN(C1)C(=O)OC(C)(C)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H19NO5/c1-10(2,3)17-9(14)12-6-5-11(15,7-12)8(13)16-4/h15H,5-7H2,1-4H3
Formula Weight 245.1263
Chemical Name or Material 1-tert-Butyl-3-Methyl 3-Hydroxypyrrolidine-1,3-Dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.