CAS RN 942190-61-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-tert-Butyl-3-Methyl 3-Hydroxypyrrolidine-1,3-Dicarboxylate, TRC
CAS: 942190-61-6 Molecular Formula: C11H19NO5 Molecular Weight (g/mol): 245.27 Synonym: 1-(1,1-Dimethylethyl) 3-methyl 3-hydroxy-1,3-pyrrolidinedicarboxylate IUPAC Name: 1-O-tert-butyl 3-O-methyl 3-hydroxypyrrolidine-1,3-dicarboxylate SMILES: COC(=O)C1(O)CCN(C1)C(=O)OC(C)(C)C