missing translation for 'onlineSavingsMsg'
Learn More

1-Palmitoyl-rac-glycerol, TRC

Product Code. 30009426
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009426 5 g 5g
30008205 1 g 1g
17928524 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009426 Supplier Toronto Research Chemicals Supplier No. TRCP1572505G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-44-9
Molecular Formula C19H38O4
Molecular Weight (g/mol) 330.5
Synonym Palmitin, 1-mono- (6CI, 8CI), (±)-1-Hexadecanoylglycerol, (±)-1-Monopalmitin, (±)-1-O-Hexadecanoylglycerol, 1-Glycerol hexadecanoate, 1-Glyceryl monohexadecanoate, 1-Monohexadecanoyl-rac-glycerol, 1-Monohexadecanoylglycerol, 1-Monopalmitin, 1-Monopalmitoyl-rac-glycerol, 1-O-Palmitoyl-DL-glycerol, 1-O-Palmitoylglycerol, 1-Palmitoyl-rac-glycerol, 1-Palmitoyl-rac-glycerol, 2,3-Dihydroxypropyl hexadecanoate, 2,3-Dihydroxypropyl palmitate, DL-1-Monopalmitin, Dimodan PA 90, Glycerol 1-monohexadecanoate, Glycerol 1-monopalmitate, Glycerol 1-palmitate, Glycerol 3-palmitate, Glyceryl palmitate, Hexadecanoic acid α-monoglyceride, NSC 404240, Palmitic acid α-monoglyceride, α-Monopalmitin, Hexadecanoic acid, 2,3-dihydroxypropyl ester
IUPAC Name 2,3-dihydroxypropyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(O)CO

Specifications

Quantity 5 g
InChI Formula InChI=1S/C19H38O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(22)23-17-18(21)16-20/h18,20-21H,2-17H2,1H3
Formula Weight 330.277
Chemical Name or Material 1-Palmitoyl-rac-glycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.