accessibility menu, dialog, popup

UserName

CAS RN 542-44-9

H3COOOHOH

CAS RN 542-44-9

IUPAC Name: 2,3-dihydroxypropyl hexadecanoate
Synonyms: glyceryl palmitate
Molecular Weight (g/mol): 330.51
Molecular Formula: C19H38O4
InChi Key: QHZLMUACJMDIAE-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OCC(O)CO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results