missing translation for 'onlineSavingsMsg'
Learn More

1-Octene, 99+%, Thermo Scientific Chemicals

Catalog Number 10424035
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
111-66-0
C8H16
112.21
MFCD00009548
KWKAKUADMBZCLK-UHFFFAOYSA-N
1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene
8125
CHEBI:46708
oct-1-ene
CCCCCCC=C
111-66-0
C8H16
112.21
MFCD00009548
KWKAKUADMBZCLK-UHFFFAOYSA-N
1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene
8125
CHEBI:46708
oct-1-ene
CCCCCCC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point -102.0°C
Density 0.7150g/mL
Boiling Point 121.0°C to 123.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)5CH=CH2
Refractive Index 1.4080 to 1.4100
Quantity 1 L
Beilstein 01, 221
Merck Index 15, 1766
Specific Gravity 0.715
Solubility Information Solubility in water: 4.1 mg/l (25°C). Other solubilities: soluble in alcohol, acetone, ether, petroleum and, coal-tar solvents, soluble in hydrocarbon solvents
Viscosity 0.38 mm2/s (40°C)
Formula Weight 112.21
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1-Octene, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'