missing translation for 'onlineSavingsMsg'
Learn More

1-Octene, 97+%

Product Code. p-7081246
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11489053

Brand: Thermo Scientific Alfa Aesar A11146.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-66-0 | C8H16 | 112.216 g/mol

1-Octene acts as a comonomer used in the preparation of polyethylene, especially high-density polyethylene (HDPE) and linear low-density polyethylene(LLDPE) resins. It is also used in organic synthesis, surfactants and plasticizers. Further, it is used in process regulators and viscosity adjustors. In addition to this, it is used in the preparation of linear aldehyde through the oxo synthesis (hydroformylation) to get the nonanal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octene acts as a comonomer used in the preparation of polyethylene, especially high-density polyethylene (HDPE) and linear low-density polyethylene(LLDPE) resins. It is also used in organic synthesis, surfactants and plasticizers. Further, it is used in process regulators and viscosity adjustors. In addition to this, it is used in the preparation of linear aldehyde through the oxo synthesis (hydroformylation) to get the nonanal.

Solubility
Miscible with water, ether, alcohol and acetone.

Notes
Incompatible with bromine, chlorine, acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-66-0
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00009548
InChI Key KWKAKUADMBZCLK-UHFFFAOYSA-N
Synonym 1-octene, caprylene, n-1-octene, 1-octylene, octylene, alpha-octene, octene-1, 1-caprylene, octene, alpha-octylene
PubChem CID 8125
ChEBI CHEBI:46708
IUPAC Name oct-1-ene
SMILES CCCCCCC=C

Specifications

Melting Point -102°C
Density 0.715
Boiling Point 122°C to 123°C
Flash Point 21°C (70°F)
Odor Petroleum-like
Refractive Index 1.409
Quantity 500 mL
UN Number UN3295
Beilstein 1734497
Merck Index 14,1764
Solubility Information Miscible with water,ether,alcohol and acetone.
Formula Weight 112.22
Percent Purity ≥97%
Chemical Name or Material 1-Octene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Octene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contains : 1-Octene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.