missing translation for 'onlineSavingsMsg'
Learn More

1-o-Tolueneazo-2-naphthol, TRC

Product Code. 30132920
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132920 5 g 5g
17973725 50 mg 50mg
17950875 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132920 Supplier Toronto Research Chemicals Supplier No. TRCT5360805G
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2646-17-5
Molecular Formula C17 H14 N2 O
Molecular Weight (g/mol) 262.31
Synonym 1-[2-(2-Methylphenyl)diazenyl]-2-naphthalenol (ACI), 2-Naphthalenol, 1-[(2-methylphenyl)azo]- (9CI), C.I. Solvent Orange 2 (7CI, 8CI), Oil Orange SS (6CI), 1-[(2-Methylphenyl)azo]-2-naphthol, 1-o-Tolueneazo-2-naphthol, 1-o-Tolylazo-2-naphthol, A.F. Orange No. 2, C.I. 12100, Dolkwal Orange SS, Ext D and C Orange No. 4, Fat Orange II, Fat Orange RR, FD and C Orange No. 2, Hexacol Oil Orange SS, Japan Orange 403, Lacquer Orange V, NSC 58044, Oil Orange 204, Oil Orange OPEL, Oil Orange TX, Orange OT, Orange SS, Orange TX, Organol Orange 2R, Solvent Orange 2, T 0553
IUPAC Name 1-[(E)-(2-methylphenyl)diazenyl]naphthalen-2-ol
SMILES Cc1ccccc1N=Nc2c(O)ccc3ccccc23

Specifications

Quantity 5 g
InChI Formula InChI=1S/C17H14N2O/c1-12-6-2-5-9-15(12)18-19-17-14-8-4-3-7-13(14)10-11-16(17)20/h2-11,20H,1H3/b19-18+
Formula Weight 262.1106
Percent Purity >95%
Chemical Name or Material 1-o-Tolueneazo-2-naphthol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.