accessibility menu, dialog, popup

UserName

CH3HNNO(Z)

CAS RN 2646-17-5

IUPAC Name: (1Z)-1-[2-(2-methylphenyl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
Synonyms: (1Z)-1-[2-(2-methylphenyl)hydrazin-1-ylidene]naphthalen-2-one
Molecular Weight (g/mol): 262.31
Molecular Formula: C17H14N2O
InChi Key: YARMASSTSUTDQJ-ZPHPHTNESA-N
SMILES: CC1=CC=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results