missing translation for 'onlineSavingsMsg'
Learn More

1-Nonen-3-ol, 98%

Product Code. 11357036
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11357036

Brand: Thermo Scientific Alfa Aesar L06889.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 21964-44-3 | C9H18O | 142.242 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21964-44-3
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00021953
InChI Key DWUPJMHAPOQKGJ-UHFFFAOYSA-N
Synonym 1-nonen-3-ol, hexylvinylcarbinol, 1-vinylheptanol, hexyl vinyl carbinol, delta1-nonen-3-ol, nonene-1-ol-3, 1-nonene-3-ol, 3-hydroxy-1-nonene, 1-nonen-3 ol, acmc-1bhje
PubChem CID 89560
IUPAC Name non-1-en-3-ol
SMILES CCCCCCC(C=C)O

Specifications

Density 0.84
Boiling Point 192°C to 194°C
Flash Point 84°C (183°F)
Refractive Index 1.442
Quantity 5 g
Beilstein 1745976
Formula Weight 142.24
Percent Purity 98%
Chemical Name or Material 1-Nonen-3-ol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.