accessibility menu, dialog, popup

UserName

CAS RN 21964-44-3

H3COHCH2

CAS RN 21964-44-3

IUPAC Name: non-1-en-3-ol
Synonyms: 1-nonen-3-ol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: DWUPJMHAPOQKGJ-UHFFFAOYNA-N
SMILES: CCCCCCC(O)C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results