missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthylmagnesium bromide, 0.5M in MeTHF

Product Code. 15458037
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458037 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15458037 Supplier Thermo Scientific Alfa Aesar Supplier No. H51159.AD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 703-55-9 | C10H7BrMg | 231.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 703-55-9
Molecular Formula C10H7BrMg
Molecular Weight (g/mol) 231.38
MDL Number MFCD00015768
InChI Key VCBWOINKHDKRAF-UHFFFAOYSA-M
Synonym 1-naphthylmagnesium bromide, naphthylmagnesium bromide, naphthyl magnesium bromide, 1-naphthyl magnesium bromide, 1-naphthyl-magnesium bromide, 1-naphthalenylmagnesium bromide, magnesium,bromo-1-naphthalenyl, pziigumposvmsd-uhfffaoysa-m, 1-naphthylmagnesium bromide solution, bromo naphthalen-1-yl magnesium, grignard reagent
PubChem CID 11831277
IUPAC Name magnesium;1H-naphthalen-1-ide;bromide
SMILES [Mg++].[Br-].C1=CC=C2[C-]=CC=CC2=C1

Specifications

Quantity 50 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Formula Weight 231.37
Concentration or Composition (by Analyte or Components) 0.5M in MeTHF
Chemical Name or Material 1-Naphthylmagnesium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.