missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 1-Naphthyl PP1

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective inhibitor of src family kinases v-Src and c-Fyn as well as the tyrosine kinase c-Abl. (IC50 values are 1.0, 0.6, 0.6, 18 and 22 μM for v-Src, c-Fyn, c-Abl, CDK2 and CAMK II respectively). Preferentially inhibits mutant over wild-type kinases (IC50 values are 1.5 vs 1000 nM for I338G v-src and v-src respectively).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 317.39 |
| Purity | >99% |
| Molecular Weight (g/mol) | 317.40 |
| Chemical Name or Material | 1-Naphthyl PP1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction