CAS RN 221243-82-9
CAS RN 221243-82-9
IUPAC Name:
1-tert-butyl-3-(naphthalen-1-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Synonyms:
1-tert-butyl-3-(naphthalen-1-yl)pyrazolo[3,4-d]pyrimidin-4-amine
Molecular Weight (g/mol):
317.4
Molecular Formula:
C19H19N5
InChi Key:
XSHQBIXMLULFEV-UHFFFAOYSA-N
SMILES:
CC(C)(C)N1N=C(C2=C(N)N=CN=C12)C1=C2C=CC=CC2=CC=C1
1
–
2
of
2
results
1
Tocris Bioscience™ 1-Naphthyl PP1
CAS: 221243-82-9 Molecular Formula: C19H19N5
2
1-Naphthyl PP1, MedChemExpress
MedChemExpress 1-Naphthyl PP1 (1-NA-PP 1) is a selective inhibitor of src family kinases. 1-Naphthyl PP1 inhibits v-Src and c-Fyn, c-Abl, CDK2 and CAMK II with IC50s of 1.0, 0.6, 0.6, 18 and 22 μM, respectively.