missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-4-methylpyridinium tetrafluoroborate, 99%

Product Code. 11446180
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
This item is not returnable. View return policy

Product Code. 11446180

Brand: Thermo Scientific Alfa Aesar H27100.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 343952-33-0 | C10H16BF4N | 237.049 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 343952-33-0
Molecular Formula C10H16BF4N
Molecular Weight (g/mol) 237.049
MDL Number MFCD03095460
InChI Key VISYYHYJMCAKAF-UHFFFAOYSA-N
Synonym 1-butyl-4-methylpyridinium tetrafluoroborate, 1-butyl-4-methylpyridin-1-ium tetrafluoroborate, acmc-1adzo, dsstox_cid_29131, dsstox_rid_83350, dsstox_gsid_49275, 1-butyl-4-picolinium tetrafluoroborate, pyridinium, 1-butyl-4-methyl-, tetrafluoroborate 1-, 1-butyl-4-methylpyridinium tetrafluoroborate t
PubChem CID 11492406
IUPAC Name 1-butyl-4-methylpyridin-1-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CCCC[N+]1=CC=C(C=C1)C

Specifications

Density 1.2
Refractive Index 1.4
Quantity 5 g
Formula Weight 237.06
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-4-methylpyridinium tetrafluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.