CAS RN 343952-33-0
CAS RN 343952-33-0
IUPAC Name:
1-butyl-4-methylpyridin-1-ium; tetrafluoroboranuide
Synonyms:
1-butyl-4-methylpyridin-1-ium tetrafluoroborate
Molecular Weight (g/mol):
237.05
Molecular Formula:
C10H16BF4N
InChi Key:
VISYYHYJMCAKAF-UHFFFAOYSA-N
SMILES:
F[B-](F)(F)F.CCCC[N+]1=CC=C(C)C=C1
1
–
1
of
1
results
1
1-n-Butyl-4-methylpyridinium tetrafluoroborate, 99%
CAS: 343952-33-0 Molecular Formula: C10H16BF4N Molecular Weight (g/mol): 237.049 MDL Number: MFCD03095460 InChI Key: VISYYHYJMCAKAF-UHFFFAOYSA-N Synonym: 1-butyl-4-methylpyridinium tetrafluoroborate,1-butyl-4-methylpyridin-1-ium tetrafluoroborate,acmc-1adzo,dsstox_cid_29131,dsstox_rid_83350,dsstox_gsid_49275,1-butyl-4-picolinium tetrafluoroborate,pyridinium, 1-butyl-4-methyl-, tetrafluoroborate 1-,1-butyl-4-methylpyridinium tetrafluoroborate t PubChem CID: 11492406 IUPAC Name: 1-butyl-4-methylpyridin-1-ium;tetrafluoroborate SMILES: [B-](F)(F)(F)F.CCCC[N+]1=CC=C(C=C1)C