missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-n-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
50 g
Description
1-n-Butyl-1-methylpyrrolidinium bis (trifluoromethylsulfonyl) imide is useful for electrodeposition. It is also used in energy storage in lithium batteries and in super capacitors. Further, it is employed in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-n-Butyl-1-methylpyrrolidinium bis (trifluoromethylsulfonyl) imide is useful for electrodeposition. It is also used in energy storage in lithium batteries and in super capacitors. Further, it is employed in organic synthesis.
Solubility
Miscible with methanol, acetonitrile, acetone, dichloromethane and dimethyl sulfoxide. Immiscible with water and diethyl ether.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents. Store in a cool place.
1-n-Butyl-1-methylpyrrolidinium bis (trifluoromethylsulfonyl) imide is useful for electrodeposition. It is also used in energy storage in lithium batteries and in super capacitors. Further, it is employed in organic synthesis.
Solubility
Miscible with methanol, acetonitrile, acetone, dichloromethane and dimethyl sulfoxide. Immiscible with water and diethyl ether.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents. Store in a cool place.
Specifications
Quantity | 5 g |
Solubility Information | Miscible with methanol,acetonitrile,acetone,dichloromethane and dimethyl sulfoxide. Immiscible with water and diethyl ether. |
Formula Weight | 422.42 |
Percent Purity | 98% |
Chemical Name or Material | 1-n-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'