missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide, 98%

Product Code. 11426330
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426330 5 g 5g
11436330 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11426330 Supplier Thermo Scientific Alfa Aesar Supplier No. H27177.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 223437-11-4 | C11H20F6N2O4S2 | 422.40 g/mol

1-n-Butyl-1-methylpyrrolidinium bis (trifluoromethylsulfonyl) imide is useful for electrodeposition. It is also used in energy storage in lithium batteries and in super capacitors. Further, it is employed in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-n-Butyl-1-methylpyrrolidinium bis (trifluoromethylsulfonyl) imide is useful for electrodeposition. It is also used in energy storage in lithium batteries and in super capacitors. Further, it is employed in organic synthesis.

Solubility
Miscible with methanol, acetonitrile, acetone, dichloromethane and dimethyl sulfoxide. Immiscible with water and diethyl ether.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 223437-11-4
Molecular Formula C11H20F6N2O4S2
Molecular Weight (g/mol) 422.40
MDL Number MFCD07784447
InChI Key HSLXOARVFIWOQF-UHFFFAOYSA-N
Synonym 1-butyl-1-methylpyrrolidinium bis trifluoromethylsulfonyl imide, unii-fd74dkk1wh, fd74dkk1wh, 1-butyl-1-methylpyrrolidinium bis trifluoromethanesulfonyl imide, 1-butyl-1-methylpyrrolidin-1-ium bis trifluoromethyl sulfonyl amide, 1-butyl-1-methylpyrrolidin-1-ium; bistriflylimide anion, dsstox_cid_27944, dsstox_rid_82696, dsstox_gsid_47968, pyrrolidinium, 1-butyl-1-methyl-, salt with 1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide 1:1
PubChem CID 11048104
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1-butyl-1-methylpyrrolidin-1-ium
SMILES CCCC[N+]1(C)CCCC1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 5 g
Solubility Information Miscible with methanol,acetonitrile,acetone,dichloromethane and dimethyl sulfoxide. Immiscible with water and diethyl ether.
Formula Weight 422.42
Percent Purity 98%
Chemical Name or Material 1-n-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.