missing translation for 'onlineSavingsMsg'
Learn More

1-N-BOC-Aminocyclopentanecarboxylic acid, 98%, Thermo Scientific™

Product Code. 10324212 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10324212 5 g Glass Bottle 5g
10129941 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10324212 Supplier Thermo Scientific Supplier No. 367340050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Identificadores de productos químicos

CAS 35264-09-6
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD01076126
InChI Key YBZCSKVLXBOFSL-UHFFFAOYSA-N
Synonym boc-cycloleucine, 1-boc-amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, n-boc-cycloleucine, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid
PubChem CID 2734645
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O

Especificaciones

Melting Point 130.0°C to 135.0°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in methanol,insoluble in toluene
Quantity 5 g
Formula Weight 229.28
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-N-BOC-Aminocyclopentanecarboxylic acid
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.