accessibility menu, dialog, popup

UserName

CH3H3CCH3OONHOO

CAS RN 35264-09-6

IUPAC Name: 1-{[(tert-butoxy)carbonyl]amino}cyclopentane-1-carboxylate
Synonyms: 1-[(tert-butoxycarbonyl)amino]cyclopentane-1-carboxylate
Molecular Weight (g/mol): 228.27
Molecular Formula: C11H18NO4
InChi Key: YBZCSKVLXBOFSL-UHFFFAOYSA-M
SMILES: CC(C)(C)OC(=O)NC1(CCCC1)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)