missing translation for 'onlineSavingsMsg'
Learn More

1-N-BOC-Aminocyclopentanecarboxylic acid, 98%, Thermo Scientific™

Product Code. 10129941 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10129941 1 g Glass Bottle 1g
10324212 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10129941 Supplier Thermo Scientific Acros Supplier No. 367340010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 35264-09-6
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD01076126
InChI Key YBZCSKVLXBOFSL-UHFFFAOYSA-N
Synonym boc-cycloleucine, 1-boc-amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, n-boc-cycloleucine, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid
PubChem CID 2734645
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O

Specifications

Melting Point 130.0°C to 135.0°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in methanol,insoluble in toluene
Quantity 1 g
Formula Weight 229.28
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-N-BOC-Aminocyclopentanecarboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.