Learn More
1-Methylimidazole-2-carboxaldehyde, 98%, Thermo Scientific Chemicals
POA - £110.00
Chemical Identifiers
CAS | 13750-81-7 |
---|---|
Molecular Formula | C5H6N2O |
Molecular Weight (g/mol) | 110.12 |
MDL Number | MFCD01321308 |
InChI Key | UEBFLTZXUXZPJO-UHFFFAOYSA-N |
Synonym | 1-methyl-1h-imidazole-2-carbaldehyde, 1-methyl-2-imidazolecarboxaldehyde, 1-methylimidazole-2-carboxaldehyde, 1h-imidazole-2-carboxaldehyde, 1-methyl, 2-formyl-1-methylimidazole, imidazole-2-carboxaldehyde, 1-methyl, pubchem7614, zlchem 1231, 2-formyl-l-methylimidazole, acmc-209ce6 |
PubChem CID | 139575 |
IUPAC Name | 1-methylimidazole-2-carbaldehyde |
SMILES | CN1C=CN=C1C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15441368
|
Thermo Scientific Alfa Aesar
H55201.MD |
250 mg | |||||||
15421368
|
Thermo Scientific Alfa Aesar
H55201.03 |
1 g |
£34.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15431368
|
Thermo Scientific Alfa Aesar
H55201.06 |
5 g |
£110.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in water.
Notes
Air sensitive. Store in a cool, dry place. Store in a tightly closed container. Store under nitrogen.
Chemical Identifiers
13750-81-7 | |
110.12 | |
UEBFLTZXUXZPJO-UHFFFAOYSA-N | |
139575 | |
CN1C=CN=C1C=O |
C5H6N2O | |
MFCD01321308 | |
1-methyl-1h-imidazole-2-carbaldehyde, 1-methyl-2-imidazolecarboxaldehyde, 1-methylimidazole-2-carboxaldehyde, 1h-imidazole-2-carboxaldehyde, 1-methyl, 2-formyl-1-methylimidazole, imidazole-2-carboxaldehyde, 1-methyl, pubchem7614, zlchem 1231, 2-formyl-l-methylimidazole, acmc-209ce6 | |
1-methylimidazole-2-carbaldehyde |
Specifications
13750-81-7 | |
70°C to 74°C (1 mmHg) | |
C5H6N2O | |
Air Sensitive | |
Soluble in water. | |
CN1C=CN=C1C=O | |
110.12 | |
110.12 | |
1-Methylimidazole-2-carboxaldehyde |
36°C to 39°C | |
102°C (216°F) | |
MFCD01321308 | |
1-methyl-1h-imidazole-2-carbaldehyde, 1-methyl-2-imidazolecarboxaldehyde, 1-methylimidazole-2-carboxaldehyde, 1h-imidazole-2-carboxaldehyde, 1-methyl, 2-formyl-1-methylimidazole, imidazole-2-carboxaldehyde, 1-methyl, pubchem7614, zlchem 1231, 2-formyl-l-methylimidazole, acmc-209ce6 | |
UEBFLTZXUXZPJO-UHFFFAOYSA-N | |
1-methylimidazole-2-carbaldehyde | |
139575 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only