missing translation for 'onlineSavingsMsg'
Learn More

1-Methylimidazole-2-carboxaldehyde, 98%

Product Code. 15431368
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431368 5 g 5g
15441368 250 mg 250mg
15421368 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15431368 Supplier Thermo Scientific Alfa Aesar Supplier No. H55201.06
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13750-81-7 | C5H6N2O | 110.12 g/mol

It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water.

Notes
Air sensitive. Store in a cool, dry place. Store in a tightly closed container. Store under nitrogen.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13750-81-7
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD01321308
InChI Key UEBFLTZXUXZPJO-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-2-carbaldehyde, 1-methyl-2-imidazolecarboxaldehyde, 1-methylimidazole-2-carboxaldehyde, 1h-imidazole-2-carboxaldehyde, 1-methyl, 2-formyl-1-methylimidazole, imidazole-2-carboxaldehyde, 1-methyl, pubchem7614, zlchem 1231, 2-formyl-l-methylimidazole, acmc-209ce6
PubChem CID 139575
IUPAC Name 1-methylimidazole-2-carbaldehyde
SMILES CN1C=CN=C1C=O

Specifications

Melting Point 36°C to 39°C
Boiling Point 70°C to 74°C (1 mmHg)
Flash Point 102°C (216°F)
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 110.12
Percent Purity 98%
Chemical Name or Material 1-Methylimidazole-2-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.