missing translation for 'onlineSavingsMsg'
Learn More

1-Methylhomopiperazine, 97%, Thermo Scientific™

Product Code. 15491678
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
This item is not returnable. View return policy

Product Code. 15491678

Brand: Thermo Scientific Alfa Aesar H55441.AA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4318-37-0
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00059810
InChI Key FXHRAKUEZPSMLJ-UHFFFAOYSA-N
Synonym n-methylhomopiperazine, n-methyl homopiperazine, 1-methylhomopiperazine, 1h-1,4-diazepine, hexahydro-1-methyl, hexahydro-1-methyl-1h-1,4-diazepine, 1-methyl-1,4-diazaperhydroepine, methylhomopiperazine, n-methylhomopiperizine, 4-methylhomopiperazine, 1-methylhomopiperizine
PubChem CID 228349
IUPAC Name 1-methyl-1,4-diazepane
SMILES CN1CCCNCC1

Specifications

Density 0.918
Boiling Point 74°C to 75°C (35 mmHg)
Flash Point 45°C (113°F)
Refractive Index 1.477
Quantity 5 mL
UN Number UN2920
Formula Weight 114.19
Percent Purity 97%
Chemical Name or Material 1-Methylhomopiperazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.