accessibility menu, dialog, popup

UserName

CAS RN 4318-37-0

CH3NHN

CAS RN 4318-37-0

IUPAC Name: 1-methyl-1,4-diazepane
Synonyms: N-methylhomopiperazine nmethylhomopiperazine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: FXHRAKUEZPSMLJ-UHFFFAOYSA-N
SMILES: CN1CCCNCC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)