missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyridone, TRC

Product Code. 30480233
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480233 500 mg 500mg
30464559 100 mg 100mg
30475333 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480233 Supplier Toronto Research Chemicals Supplier No. TRCM156471500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-85-9
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.12
Synonym 1-Methyl-2(1H)-pyridinone, 1-Methyl-1,2-dihydro-2-pyridinone, 1-Methyl-1H-pyridin-2-one, 1-Methyl-2(1H)-pyridone, 1-Methyl-2-oxopyridine, 1-Methyl-2-pyridinone, 1-Methylpyridin-1-ium-2-olate, N-Methyl-2-pyridone, NSC 9383
IUPAC Name 1-methylpyridin-2-one
SMILES CN1C=CC=CC1=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H7NO/c1-7-5-3-2-4-6(7)8/h2-5H,1H3
Formula Weight 109.0528
Chemical Name or Material 1-Methyl-2-pyridone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.