accessibility menu, dialog, popup

UserName

CH3NO

CAS RN 694-85-9

IUPAC Name: 1-methyl-1,2-dihydropyridin-2-one
Synonyms: 2(1H)-pyridinone, 1-methyl-
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: DVVGIUUJYPYENY-UHFFFAOYSA-N
SMILES: CN1C=CC=CC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)