missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-6-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-3484809
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10739415 250 mg Amber Glass Bottle 250mg
10517294 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10739415 Supplier Thermo Scientific Maybridge Supplier No. CC44201CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 202745-73-1
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD08690250
InChI Key DJQOGNYSBOIJKE-UHFFFAOYSA-N
PubChem CID 19819261
IUPAC Name 1-methylindole-6-carboxylic acid
SMILES CN1C=CC2=C1C=C(C=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-methyl-1h-indole-6-carboxylic acid, 1h-indole-6-carboxylic acid, 1-methyl, 1-methyl-6-indolecarboxylic acid, 6-carboxy-1-methyl-1h-indole, 1-methyl-ih-indole-6-carboxylic acid, 1h-indole-6-carboxylicacid,1-methyl, 1h-indole-6-carboxylicacid, 1-methyl
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-6-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.