accessibility menu, dialog, popup

UserName

CAS RN 202745-73-1

H3CNOOH

CAS RN 202745-73-1

IUPAC Name: 1-methyl-1H-indole-6-carboxylic acid
Synonyms: 1-methylindole-6-carboxylic acid
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: DJQOGNYSBOIJKE-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC=C(C=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results