missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy Empagliflozin, TRC

Product Code. 30134677
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134677 25 mg 25mg
17910395 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134677 Supplier Toronto Research Chemicals Supplier No. TRCM26249325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1279691-36-9
Molecular Formula C24H29ClO8
Molecular Weight (g/mol) 480.94
Synonym Methyl 1-C-[4-Chloro-3-[[4-[[(3S)-tetrahydro-3-furanyl]oxy]phenyl]methyl]phenyl]-α-D-glucopyranoside;
IUPAC Name (2S,3R,4S,5S,6R)-2-[4-chloro-3-[[4-[(3S)-oxolan-3-yl]oxyphenyl]methyl]phenyl]-6-(hydroxymethyl)-2-methoxyoxane-3,4,5-triol
SMILES CO[C@]1(C2=CC=C(C(=C2)CC3=CC=C(C=C3)O[C@H]4CCOC4)Cl)[C@@H]([C@H]([C@@H]([C@@H](CO)O1)O)O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C24H29ClO8/c1-30-24(23(29)22(28)21(27)20(12-26)33-24)16-4-7-19(25)15(11-16)10-14-2-5-17(6-3-14)32-18-8-9-31-13-18/h2-7,11,18,20-23,26-29H,8-10,12-13H2,1H3/t18-,20+,21+,22-,23+,24-/m0/s1
Formula Weight 480.1551
Percent Purity >95%
Chemical Name or Material 1-Methoxy Empagliflozin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.