CAS RN 1279691-36-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Methoxy Empagliflozin, TRC
CAS: 1279691-36-9 Molecular Formula: C24H29ClO8 Molecular Weight (g/mol): 480.94 Synonym: Methyl 1-C-[4-Chloro-3-[[4-[[(3S)-tetrahydro-3-furanyl]oxy]phenyl]methyl]phenyl]-α-D-glucopyranoside; IUPAC Name: (2S,3R,4S,5S,6R)-2-[4-chloro-3-[[4-[(3S)-oxolan-3-yl]oxyphenyl]methyl]phenyl]-6-(hydroxymethyl)-2-methoxyoxane-3,4,5-triol SMILES: CO[C@]1(C2=CC=C(C(=C2)CC3=CC=C(C=C3)O[C@H]4CCOC4)Cl)[C@@H]([C@H]([C@@H]([C@@H](CO)O1)O)O)O