missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-piperidinone Hydrochloride, TRC

Product Code. 30369201
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369201 100 mg -
30377611 50 mg 50mg
30386270 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369201 Supplier Toronto Research Chemicals Supplier No. TRCE939318100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41361-28-8
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
Synonym 1-Ethyl-3-piperidone Hydrochloride
IUPAC Name 1-ethylpiperidin-3-one hydrochloride
SMILES Cl.O=C1CN(CC)CCC1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H13NO.ClH/c1-2-8-5-3-4-7(9)6-8;/h2-6H2,1H3;1H
Formula Weight 163.08
Chemical Name or Material 1-Ethyl-3-piperidinone Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.