CAS RN 41361-28-8
IUPAC Name:
hydrogen 1-ethylpiperidin-3-one chloride
Synonyms:
hydrogen 1-ethylpiperidin-3-one chloride
Molecular Weight (g/mol):
163.65
Molecular Formula:
C7H14ClNO
InChi Key:
JLBJGYHSFYAUDJ-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CCN1CCCC(=O)C1
1
–
1
of
1
results
1
1-Ethyl-3-piperidinone Hydrochloride, TRC
CAS: 41361-28-8 Molecular Formula: C7H16ClNO2 Molecular Weight (g/mol): 181.66 Synonym: 1-Ethyl-3-piperidone Hydrochloride IUPAC Name: 1-ethylpiperidin-3-one hydrochloride SMILES: Cl.O=C1CN(CC)CCC1