missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-fluoro-2-nitrobenzene, 98+%, Thermo Scientific™

Product Code. 11470233
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470233 25 g 25g
11450233 1 g 1g
11460233 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470233 Supplier Thermo Scientific Alfa Aesar Supplier No. A11400.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 345-17-5
Molecular Formula C6H3ClFNO2
Molecular Weight (g/mol) 175.54
MDL Number MFCD00084853
InChI Key DVXDJQKEEKXJBW-UHFFFAOYSA-N
Synonym 2-chloro-5-fluoronitrobenzene, benzene, 1-chloro-4-fluoro-2-nitro, pubchem1049, ksc497k7f, 1-chloro-4-fluoronitrobenzene, 2-chloro-5-fiuoronitrobenzene, 2-chloro-5-fluoro-nitrobenzene, 2-chloro-5-fluoronitro-benzene, ablock ab-13-8500, 1-chloro-4-fluoro-2-nitro-benzene
PubChem CID 67659
IUPAC Name 1-chloro-4-fluoro-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=CC=C1Cl

Specifications

Melting Point 37°C to 39°C
Boiling Point 237°C to 239°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 25 g
UN Number UN2811
Beilstein 2094191
Formula Weight 175.55
Percent Purity ≥98%
Chemical Name or Material 1-Chloro-4-fluoro-2-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.