CAS RN 345-17-5
CAS RN 345-17-5
IUPAC Name:
1-chloro-4-fluoro-2-nitrobenzene
Synonyms:
1-chloro-4-fluoro-2-nitrobenzene
Molecular Weight (g/mol):
175.54
Molecular Formula:
C6H3ClFNO2
InChi Key:
DVXDJQKEEKXJBW-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(F)=CC=C1Cl
1
–
1
of
1
results
1
1-Chloro-4-fluoro-2-nitrobenzene, 98+%, Thermo Scientific™
CAS: 345-17-5 Molecular Formula: C6H3ClFNO2 Molecular Weight (g/mol): 175.54 MDL Number: MFCD00084853 InChI Key: DVXDJQKEEKXJBW-UHFFFAOYSA-N Synonym: 2-chloro-5-fluoronitrobenzene,benzene, 1-chloro-4-fluoro-2-nitro,pubchem1049,ksc497k7f,1-chloro-4-fluoronitrobenzene,2-chloro-5-fiuoronitrobenzene,2-chloro-5-fluoro-nitrobenzene,2-chloro-5-fluoronitro-benzene,ablock ab-13-8500,1-chloro-4-fluoro-2-nitro-benzene PubChem CID: 67659 IUPAC Name: 1-chloro-4-fluoro-2-nitrobenzene SMILES: [O-][N+](=O)C1=CC(F)=CC=C1Cl