missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-methyl-2-phenylpropane, 98%, Thermo Scientific™

Product Code. 10265360 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265360 100 mL Glass bottle 100mL
10205320 5 mL Glass bottle 5mL
10647674 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265360 Supplier Thermo Scientific Acros Supplier No. 124341000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 515-40-2
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.66
MDL Number MFCD00000940
InChI Key DNXXUUPUQXSUFH-UHFFFAOYSA-N
Synonym neophyl chloride, 1-chloro-2-methyl-2-phenylpropane, 1-chloro-2-methylpropan-2-yl benzene, benzene, 2-chloro-1,1-dimethylethyl, 2-methyl-2-phenylpropyl chloride, neophylchloride, 2-chloro-1,1-dimethylethyl benzene, unii-of6e0410nf, beta-chloro-tert-butylbenzene, 2-chloromethyl-2-phenylpropane
PubChem CID 68191
IUPAC Name (1-chloro-2-methylpropan-2-yl)benzene
SMILES CC(C)(CCl)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 95°C to 100°C (10.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C(CH3)2CH2Cl
Refractive Index 1.5230 to 1.525
Quantity 100 mL
Beilstein 05,II,32
Merck Index 15,6544
Specific Gravity 1.04
Solubility Information Solubility in water: insoluble
Formula Weight 168.67
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-methyl-2-phenylpropane, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Chloro-2-methyl-2-phenylpropane
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.