accessibility menu, dialog, popup

UserName

H3CCH3Cl

CAS RN 515-40-2

IUPAC Name: (1-chloro-2-methylpropan-2-yl)benzene
Synonyms: neophyl chloride
Molecular Weight (g/mol): 168.66
Molecular Formula: C10H13Cl
InChi Key: DNXXUUPUQXSUFH-UHFFFAOYSA-N
SMILES: CC(C)(CCl)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)