Learn More
Integra Biosciences™ 1-Channel Adapter
For use with VACUBOY Hand Operator.
£13.85 - £34.00
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11777312
|
Integra Biosciences™
159027 |
1-channel adaptor with ejecter for GripTips |
£31.95
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
10090272
|
Integra Biosciences™
155522 |
1-channel stainless steel adapter, 150 mm |
£30.75
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
10319341
|
Integra Biosciences™
155502 |
1-channel stainless steel adapter, 40 mm |
£34.00
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
12964417
|
Integra Biosciences™
159023 |
1-channel adapter for disposable tips (Pk of 5) |
£13.85
Pack of 5 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
One-channel adaptor for use with VACUBOY hand operator.The VACUBOY hand operator can be used with VACUSAFE comfort vacuum system. Enables liquid residues to be disposed of reliably, precisely and conveniently:
- With a precisely dosed volume
- Without loss of drops
- Without risk of contamination
- Without causing tiredness
- Energy saving, the manual operator - the heart of the system - can be operated as a single or 8 channel aspirator
- All parts in contact with fluids are autoclavable
Specifications
VACUBOY Hand Operator |