missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-fluoro-2-nitrobenzene, 98%, Thermo Scientific™

Product Code. 11358286
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358286 250 mg 250mg
11338286 1 g 1g
11348286 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11358286 Supplier Thermo Scientific Alfa Aesar Supplier No. L15503.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-09-3
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 219.997
MDL Number MFCD00055530
InChI Key XRXNWKIKQFEOGO-UHFFFAOYSA-N
Synonym 2-bromo-5-fluoronitrobenzene, benzene, 1-bromo-4-fluoro-2-nitro, 2-bromo-5-fluoro-nitrobenzene, pubchem1044, acmc-1ajt6, ksc494a1j, 2-nitro-4-fluoro-bromobenzene, xrxnwkikqfeogo-uhfffaoysa, 2-bromo-5-fluoro nitro benzene, 1-bromo-2-nitro-4-fluorobenzene
PubChem CID 2773383
IUPAC Name 1-bromo-4-fluoro-2-nitrobenzene
SMILES C1=CC(=C(C=C1F)[N+](=O)[O-])Br

Specifications

Melting Point 39°C to 42°C
Boiling Point 148°C to 150°C (35 mmHg)
Quantity 250 mg
UN Number UN2811
Beilstein 2364227
Formula Weight 220.01
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-fluoro-2-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.