accessibility menu, dialog, popup

UserName

ONOFBr

CAS RN 446-09-3

IUPAC Name: 1-bromo-4-fluoro-2-nitrobenzene
Synonyms: 1-bromo-4-fluoro-2-nitrobenzene
Molecular Weight (g/mol): 220
Molecular Formula: C6H3BrFNO2
InChi Key: XRXNWKIKQFEOGO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=CC=C1Br
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results