missing translation for 'onlineSavingsMsg'
Learn More

1-Benzofuran-5-carbonitrile, 97%, Thermo Scientific™

Product Code. 10311203
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10311203 10 g Amber Glass Bottle 10g
10791084 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10311203 Supplier Thermo Scientific Maybridge Supplier No. CC07116EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 79002-39-4
Molecular Formula C9H5NO
Molecular Weight (g/mol) 143.15
MDL Number MFCD05664402
InChI Key SXFQAFFSEZRQCJ-UHFFFAOYSA-N
Synonym benzofuran-5-carbonitrile, 5-benzofurancarbonitrile, 1-benzofurane-5-carbonitrile, 5-cyano-benzofuran, cyanobenzofuran, 5-cyanobenzo b furan, pubchem9873, benzo b furan-5-carbonitrile, 1-benzo b furan-5-carbonitrile, buttpark 182\50-55
PubChem CID 2795184
IUPAC Name 1-benzofuran-5-carbonitrile
SMILES N#CC1=CC=C2OC=CC2=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 143.14
Percent Purity 97%
Chemical Name or Material 1-Benzofuran-5-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.