accessibility menu, dialog, popup

UserName

ON

CAS RN 79002-39-4

IUPAC Name: 1-benzofuran-5-carbonitrile
Synonyms: 1-benzofuran-5-carbonitrile
Molecular Weight (g/mol): 143.15
Molecular Formula: C9H5NO
InChi Key: SXFQAFFSEZRQCJ-UHFFFAOYSA-N
SMILES: N#CC1=CC=C2OC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results