missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Adamantanol, 99%, Thermo Scientific Chemicals
£15.60 - £147.00
Chemical Identifiers
CAS | 768-95-6 |
---|---|
Molecular Formula | C10H16O |
Molecular Weight (g/mol) | 152.24 |
MDL Number | MFCD00074729 |
InChI Key | VLLNJDMHDJRNFK-UHFFFAOYSA-N |
Synonym | 1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol |
PubChem CID | 64152 |
IUPAC Name | adamantan-1-ol |
SMILES | C1C2CC3CC1CC(C2)(C3)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10397660
|
Thermo Scientific Acros
157100050 |
5 g | Glass bottle |
£15.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10193250
|
Thermo Scientific Acros
157100250 |
25 g | Glass bottle |
£42.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10485165
|
Thermo Scientific Acros
157101000 |
100 g | Glass bottle |
£147.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
768-95-6 | |
152.24 | |
VLLNJDMHDJRNFK-UHFFFAOYSA-N | |
64152 | |
C1C2CC3CC1CC(C2)(C3)O |
C10H16O | |
MFCD00074729 | |
1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol | |
adamantan-1-ol |
Specifications
768-95-6 | |
100.0 | |
White | |
98.5% min. (GC) | |
MFCD00074729 | |
Solubility in water: insoluble in water. Other solubilities: soluble in ethanol,diethyl ether,methylene,chloride and acetone | |
C1C2CC3CC1CC(C2)(C3)O | |
152.24 | |
152.24 | |
Crystalline Powder or Needles |
98.5 | |
>240.0°C | |
Authentic | |
C10H16O | |
1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol | |
VLLNJDMHDJRNFK-UHFFFAOYSA-N | |
adamantan-1-ol | |
64152 | |
99% | |
1-Adamantanol |
Safety and Handling
missing translation for 'einecsNumber' : 212-202-8
RUO – Research Use Only