missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanol, 99%

Product Code. 10397660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10397660 5 g Glass bottle 5g
10193250 25 g Glass bottle 25g
10485165 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10397660 Supplier Thermo Scientific Acros Supplier No. 157100050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 768-95-6 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-95-6
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00074729
InChI Key VLLNJDMHDJRNFK-UHFFFAOYSA-N
Synonym 1-adamantanol, 1-hydroxyadamantane, 1-adamantol, adamantanol, tricyclo 3.3.1.1 3,7 decan-1-ol, tricyclo 3.3.1.1∼3,7∼ decan-1-ol, 1-adamantanol 1-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3r,5s,7s-adamantan-1-ol
PubChem CID 64152
IUPAC Name adamantan-1-ol
SMILES C1C2CC3CC1CC(C2)(C3)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >240.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol,diethyl ether,methylene,chloride and acetone
Formula Weight 152.24
Percent Purity 99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 1-Adamantanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.