missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-(isobutylamino)piperidine, 98%, Thermo Scientific™

Product Code. 11404720
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11404720

Brand: Thermo Scientific Alfa Aesar H50763.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902836-84-4
Molecular Formula C11H22N2O
Molecular Weight (g/mol) 198.31
MDL Number MFCD08060973
InChI Key YQFYWSRHLRHQDZ-UHFFFAOYSA-N
Synonym 1-acetyl-4-isobutylamino piperidine, 1-4-isobutylamino-piperidin-1-yl-ethanone, 1-4-2-methylpropyl amino piperidin-1-yl ethanone, 1-4-isobutylamino piperidin-1-yl ethanone, 1-acetyl-4-2-methylpropyl amino piperidine
PubChem CID 24208818
IUPAC Name 1-[4-(2-methylpropylamino)piperidin-1-yl]ethanone
SMILES CC(C)CNC1CCN(CC1)C(=O)C

Specifications

Quantity 1 g
Formula Weight 198.31
Percent Purity 98%
Chemical Name or Material 1-Acetyl-4-(isobutylamino)piperidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.