accessibility menu, dialog, popup

UserName

CAS RN 902836-84-4

H3CCH3NHNOH3C

CAS RN 902836-84-4

IUPAC Name: 1-{4-[(2-methylpropyl)amino]piperidin-1-yl}ethan-1-one
Synonyms: 1-{4-[(2-methylpropyl)amino]piperidin-1-yl}ethanone
Molecular Weight (g/mol): 198.31
Molecular Formula: C11H22N2O
InChi Key: YQFYWSRHLRHQDZ-UHFFFAOYSA-N
SMILES: CC(C)CNC1CCN(CC1)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results