missing translation for 'onlineSavingsMsg'
Learn More

1-(9-Fluorenylmethyl)piperidine, TRC

Product Code. 30401539
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401539 500 mg
30407234 100 mg
30401155 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401539 Supplier Toronto Research Chemicals Supplier No. TRCF532445500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35661-58-6
Molecular Formula C10 H15 N O6 S
Molecular Weight (g/mol) 277.294
Synonym 1-(9H-Fluoren-9-ylmethyl)piperidine
IUPAC Name [4-[1-hydroxy-2-(methylamino)ethyl]-2-methoxyphenyl] hydrogen sulfate
SMILES CNCC(O)c1ccc(OS(=O)(=O)O)c(OC)c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H15NO6S/c1-11-6-8(12)7-3-4-9(10(5-7)16-2)17-18(13,14)15/h3-5,8,11-12H,6H2,1-2H3,(H,13,14,15)
Formula Weight 277.062
Chemical Name or Material 1-(9-Fluorenylmethyl)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.