CAS RN 35661-58-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(9-Fluorenylmethyl)piperidine, TRC
CAS: 35661-58-6 Molecular Formula: C10 H15 N O6 S Molecular Weight (g/mol): 277.294 Synonym: 1-(9H-Fluoren-9-ylmethyl)piperidine IUPAC Name: [4-[1-hydroxy-2-(methylamino)ethyl]-2-methoxyphenyl] hydrogen sulfate SMILES: CNCC(O)c1ccc(OS(=O)(=O)O)c(OC)c1